Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN15
Entrez Gene ID
102178033
Gene Name
claudin 15
Gene Aliases
-
Chromosome Location
Chr.25: 36373628 - 36380449 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013974836.2 XP_013830290.1 1 - 2 1799 72
XM_018040877.1 XP_017896366.1 2 - 3 1659 72
XM_018040878.1 XP_017896367.1 2 - 3 1682 72
XM_018040880.1 XP_017896369.1 1 - 2 1995 72

View Full Product Details