Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN15
Entrez Gene ID
102178033
Gene Name
claudin 15
Gene Aliases
-
Chromosome Location
Chr.25: 36373628 - 36380449 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013974836.2 XP_013830290.1 4 - 5 2163 97
XM_018040877.1 XP_017896366.1 5 - 6 2023 97
XM_018040878.1 XP_017896367.1 5 - 6 2046 97
XM_018040879.1 XP_017896368.1 4 - 5 2097 97
XM_018040880.1 XP_017896369.1 4 - 5 2390 97

View Full Product Details