Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMMECR1
Entrez Gene ID
102182574
Gene Name
Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189519: 149188 - 210091 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013976205.2 XP_013831659.1 3 - 4 623 109
GenBank mRNA JT705034.1 - - 896 109

View Full Product Details