Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DKK2
Entrez Gene ID
102188662
Gene Name
dickkopf WNT signaling pathway inhibitor 2
Gene Aliases
-
Chromosome Location
Chr.6: 18461851 - 18590124 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681333.3 XP_005681390.1 2 - 3 1095 89
GenBank mRNA KA341311.1 - 2 - 3 915 89

View Full Product Details