Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF79
Entrez Gene ID
102190046
Gene Name
zinc finger protein 79
Gene Aliases
-
Chromosome Location
Chr.11: 97707439 - 97721872 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055808.1 XP_017911297.1 2 - 3 445 77
GenBank mRNA JO438826.2 - 3 - 4 731 77
JO592986.2 - 2 - 3 436 77

View Full Product Details