Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBBP9
Entrez Gene ID
102190614
Gene Name
RB binding protein 9, serine hydrolase
Gene Aliases
-
Chromosome Location
Chr.13: 37939725 - 37947241 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688073.3 XP_005688130.1 4 - 5 389 97

View Full Product Details