Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102189789
Entrez Gene ID
102189789
Gene Name
zinc finger protein 91
Gene Aliases
ZNF234
Chromosome Location
Chr.18: 53155385 - 53191295 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013971412.2 XP_013826866.2 3 - 4 451 87

View Full Product Details