Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPRSS11D
Entrez Gene ID
102186382
Gene Name
transmembrane serine protease 11D
Gene Aliases
-
Chromosome Location
Chr.6: 84243473 - 84303504 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013964668.2 XP_013820122.1 4 - 5 436 102

View Full Product Details