Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LDLRAD3
Entrez Gene ID
102183254
Gene Name
low density lipoprotein receptor class A domain containing 3
Gene Aliases
-
Chromosome Location
Chr.15: 16220105 - 16489634 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059234.1 XP_017914723.1 3 - 4 374 52
XM_018059236.1 XP_017914725.1 3 - 4 422 52
GenBank mRNA JO426523.1 - 3 - 4 547 52

View Full Product Details