Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRP5
Entrez Gene ID
106503755
Gene Name
LDL receptor related protein 5
Gene Aliases
LRP5L
Chromosome Location
Chr.29: 46841558 - 46964043 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043250.1 XP_017898739.1 8 - 9 1929 112
XM_018043251.1 XP_017898740.1 8 - 9 1845 112

View Full Product Details