Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C3H1orf50
Entrez Gene ID
102176385
Gene Name
chromosome 3 C1orf50 homolog
Gene Aliases
-
Chromosome Location
Chr.3: 17313997 - 17323505 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678598.3 XP_005678655.3 3 - 4 336 72
XM_018042680.1 XP_017898169.1 3 - 4 327 72
GenBank mRNA JO438657.2 - 3 - 4 316 72

View Full Product Details