Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C5H12orf50
Entrez Gene ID
102169833
Gene Name
chromosome 5 C12orf50 homolog
Gene Aliases
-
Chromosome Location
Chr.5: 17580967 - 17610051 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679783.1 XP_005679840.1 12 - 13 1388 103

View Full Product Details