Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FICD
Entrez Gene ID
102180789
Gene Name
FIC domain containing
Gene Aliases
-
Chromosome Location
Chr.17: 6689918 - 6695117 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061078.1 XP_017916567.1 2 - 3 454 66
GenBank mRNA JO587061.2 - 2 - 3 443 66
JO587062.2 - 2 - 3 443 66

View Full Product Details