Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C25H16orf58
Entrez Gene ID
102181919
Gene Name
chromosome 25 C16orf58 homolog
Gene Aliases
-
Chromosome Location
Chr.25: 27398124 - 27413878 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO426112.1 - 2 - 3 255 63
JO426157.1 - 2 - 3 255 63
JO637366.2 - 3 - 4 437 63

View Full Product Details