The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018041119.1 | XP_017896608.1 | 3 - 4 | 637 | 100 |
GenBank mRNA | JO356366.2 | - | 3 - 4 | 488 | 100 |