Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL10
Entrez Gene ID
102175673
Gene Name
ADP ribosylation factor like GTPase 10
Gene Aliases
-
Chromosome Location
Chr.7: 72759928 - 72769370 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050657.1 XP_017906146.1 2 - 3 497 87
GenBank mRNA KA342474.1 - 2 - 3 416 87

View Full Product Details