Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C26H10orf90
Entrez Gene ID
102174184
Gene Name
chromosome 26 C10orf90 homolog
Gene Aliases
-
Chromosome Location
Chr.26: 5863524 - 6107092 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013975309.2 XP_013830763.2 7 - 8 2383 75

View Full Product Details