Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102182016
Entrez Gene ID
102182016
Gene Name
protocadherin-19
Gene Aliases
PCDH19
Chromosome Location
Chr.Un|NW_017215092: 18760 - 21885 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045554.1 XP_017901043.1 - 3694 70

View Full Product Details