Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM13
Entrez Gene ID
102169671
Gene Name
PR/SET domain 13
Gene Aliases
-
Chromosome Location
Chr.9: 37475145 - 37483622 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053530.1 XP_017909019.1 3 - 4 659 71

View Full Product Details