Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RDH11
Entrez Gene ID
102171883
Gene Name
retinol dehydrogenase 11
Gene Aliases
-
Chromosome Location
Chr.10: 23590584 - 23603151 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686002.3 XP_005686059.1 1 - 2 322 88
GenBank mRNA JT710415.1 - 8 - 9 1892 88

View Full Product Details