Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIEN1
Entrez Gene ID
102182627
Gene Name
migration and invasion enhancer 1
Gene Aliases
-
Chromosome Location
Chr.19: 39737973 - 39739386 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693754.3 XP_005693811.1 3 - 4 332 92
GenBank mRNA JO399989.2 - 3 - 4 322 92

View Full Product Details