Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NKAPL
Entrez Gene ID
102172222
Gene Name
NFKB activating protein like
Gene Aliases
-
Chromosome Location
Chr.23: 19500739 - 19502714 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696707.3 XP_005696764.2 1 - 1 662 91

View Full Product Details