Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGF4
Entrez Gene ID
102170206
Gene Name
fibroblast growth factor 4
Gene Aliases
-
Chromosome Location
Chr.29: 48043184 - 48045131 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042877.1 XP_017898366.1 1 - 2 341 88

View Full Product Details