Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMGT1
Entrez Gene ID
102169945
Gene Name
membrane magnesium transporter 1
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189517: 20407786 - 20420614 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044237.1 XP_017899726.1 2 - 3 152 85

View Full Product Details