Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NYNRIN
Entrez Gene ID
102177167
Gene Name
NYN domain and retroviral integrase containing
Gene Aliases
-
Chromosome Location
Chr.10: 80658794 - 80680139 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054648.1 XP_017910137.1 3 - 4 1014 69

View Full Product Details