Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LONRF1
Entrez Gene ID
102172512
Gene Name
LON peptidase N-terminal domain and ring finger 1
Gene Aliases
-
Chromosome Location
Chr.27: 21321584 - 21369063 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042095.1 XP_017897584.1 5 - 6 1456 103

View Full Product Details