Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTPBP8
Entrez Gene ID
102183545
Gene Name
GTP binding protein 8 (putative)
Gene Aliases
-
Chromosome Location
Chr.1: 57472692 - 57492927 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005674926.3 XP_005674983.2 4 - 5 704 81
XM_018066550.1 XP_017922039.1 4 - 5 718 81
GenBank mRNA JO434187.2 - 4 - 5 706 81
JO583340.1 - 2 - 3 259 81
JO583341.1 - 2 - 3 259 81
KA347662.1 - 2 - 3 431 81

View Full Product Details