Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5C1A
Entrez Gene ID
102191298
Gene Name
5'-nucleotidase, cytosolic IA
Gene Aliases
-
Chromosome Location
Chr.3: 14382892 - 14406177 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678658.3 XP_005678715.1 1 - 2 274 65
XM_018042176.1 XP_017897665.1 1 - 2 274 65
GenBank mRNA JO380396.2 - 5 - 6 1145 65
JO541856.2 - 2 - 3 244 65

View Full Product Details