Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102188567
Entrez Gene ID
102188567
Gene Name
C-type lectin domain family 2 member A
Gene Aliases
CLEC2A
Chromosome Location
Chr.5: 98478945 - 98488781 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680860.3 XP_005680917.2 2 - 3 255 126
XM_005680862.3 XP_005680919.1 2 - 3 255 126

View Full Product Details