Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COMMD6
Entrez Gene ID
102181509
Gene Name
COMM domain containing 6
Gene Aliases
-
Chromosome Location
Chr.12: 35807768 - 35828459 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687660.3 XP_005687717.1 3 - 4 293 99

View Full Product Details