Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPF3A
Entrez Gene ID
102186132
Gene Name
UPF3A regulator of nonsense mediated mRNA decay
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189962: 620356 - 643325 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044973.1 XP_017900462.1 5 - 6 699 100
GenBank mRNA JO424441.1 - 5 - 6 776 100
KA330779.1 - 5 - 6 1503 100

View Full Product Details