Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POP4
Entrez Gene ID
102183260
Gene Name
POP4 homolog, ribonuclease P/MRP subunit
Gene Aliases
-
Chromosome Location
Chr.18: 41416989 - 41426025 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692232.3 XP_005692289.1 3 - 4 508 85
GenBank mRNA JO610184.2 - 4 - 5 764 85
JO629273.2 - 3 - 4 169 85

View Full Product Details