Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A11
Entrez Gene ID
102191309
Gene Name
solute carrier family 22 member 11
Gene Aliases
-
Chromosome Location
Chr.29: 43220897 - 43235140 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699974.2 XP_005700031.2 2 - 3 503 105

View Full Product Details