Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGFBPL1
Entrez Gene ID
102184997
Gene Name
insulin like growth factor binding protein like 1
Gene Aliases
-
Chromosome Location
Chr.8: 62103330 - 62119179 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018052833.1 XP_017908322.1 1 - 2 447 97
GenBank mRNA JO649179.2 - 4 - 5 351 97
JT660623.1 - 4 - 5 414 97

View Full Product Details