Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT16
Entrez Gene ID
102181397
Gene Name
synaptotagmin 16
Gene Aliases
-
Chromosome Location
Chr.10: 28944985 - 29070803 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054082.1 XP_017909571.1 5 - 6 1676 80

View Full Product Details