Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102190927
Entrez Gene ID
102190927
Gene Name
paired immunoglobulin-like type 2 receptor alpha
Gene Aliases
PILRA
Chromosome Location
Chr.25: 37010239 - 37026385 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040730.1 XP_017896219.1 5 - 6 863 84
GenBank mRNA JO587033.2 - 5 - 6 820 84

View Full Product Details