Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPN20
Entrez Gene ID
102189552
Gene Name
protein tyrosine phosphatase non-receptor type 20
Gene Aliases
PTPN20B
Chromosome Location
Chr.28: 3207632 - 3233695 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013975687.2 XP_013831141.2 3 - 4 924 117

View Full Product Details