Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TOE1
Entrez Gene ID
102184217
Gene Name
target of EGR1, exonuclease
Gene Aliases
-
Chromosome Location
Chr.3: 20052441 - 20056226 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678537.3 XP_005678594.2 2 - 3 372 82
XM_018043596.1 XP_017899085.1 2 - 3 372 82

View Full Product Details