Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB3D
Entrez Gene ID
102187671
Gene Name
RAB3D, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.7: 95598536 - 95609743 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051108.1 XP_017906597.1 2 - 3 471 80
GenBank mRNA KA323978.1 - 2 - 3 438 80

View Full Product Details