Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7H19orf25
Entrez Gene ID
102191420
Gene Name
chromosome 7 C19orf25 homolog
Gene Aliases
-
Chromosome Location
Chr.7: 66602939 - 66607895 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682916.3 XP_005682973.1 1 - 2 362 113
GenBank mRNA JO618873.2 - 1 - 2 345 113
KA316223.1 - 1 - 2 2556 113

View Full Product Details