Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RP1L1
Entrez Gene ID
102187132
Gene Name
RP1 like 1
Gene Aliases
-
Chromosome Location
Chr.8: 8496727 - 8541965 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018052756.1 XP_017908245.1 1 - 2 604 78

View Full Product Details