Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PQLC1
Entrez Gene ID
102187638
Gene Name
PQ loop repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.24: 440211 - 481778 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039390.1 XP_017894879.1 2 - 3 367 100
GenBank mRNA JO391501.2 - 3 - 4 560 100

View Full Product Details