Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMX2
Entrez Gene ID
102190850
Gene Name
empty spiracles homeobox 2
Gene Aliases
-
Chromosome Location
Chr.26: 14142771 - 14149498 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698620.3 XP_005698677.2 2 - 3 1157 84
GenBank mRNA KA334949.1 - 2 - 3 692 84

View Full Product Details