Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCB10
Entrez Gene ID
102183293
Gene Name
ATP binding cassette subfamily B member 10
Gene Aliases
-
Chromosome Location
Chr.28: 43668421 - 43702621 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042167.1 XP_017897656.1 7 - 8 1323 73

View Full Product Details