Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ETAA1
Entrez Gene ID
102181310
Gene Name
ETAA1 activator of ATR kinase
Gene Aliases
-
Chromosome Location
Chr.11: 65542833 - 65556838 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686809.3 XP_005686866.2 1 - 2 351 62

View Full Product Details