Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTR2A
Entrez Gene ID
102182605
Gene Name
5-hydroxytryptamine receptor 2A
Gene Aliases
-
Chromosome Location
Chr.12: 69719929 - 69785086 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687443.3 XP_005687500.1 3 - 4 1346 92
GenBank mRNA JO616285.2 - 2 - 3 511 92

View Full Product Details