Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PROCR
Entrez Gene ID
102171169
Gene Name
protein C receptor
Gene Aliases
-
Chromosome Location
Chr.13: 64019438 - 64024302 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688489.3 XP_005688546.1 3 - 4 463 100
XM_013968973.2 XP_013824427.1 3 - 4 491 100
XM_013968974.2 XP_013824428.1 2 - 3 510 100
XM_018057757.1 XP_017913246.1 2 - 3 510 100
GenBank mRNA JO350568.2 - 3 - 4 414 100

View Full Product Details