Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHMP4B
Entrez Gene ID
102173581
Gene Name
charged multivesicular body protein 4B
Gene Aliases
-
Chromosome Location
Chr.13: 62814613 - 62861235 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057730.1 XP_017913219.1 1 - 2 352 79
GenBank mRNA JO579103.1 - 4 - 5 747 79
KA324300.1 - 1 - 2 417 79

View Full Product Details