Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD45
Entrez Gene ID
102176351
Gene Name
ankyrin repeat domain 45
Gene Aliases
-
Chromosome Location
Chr.16: 53568647 - 53618369 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690886.3 XP_005690943.1 5 - 6 826 102
GenBank mRNA JO439929.2 - - 953 102
JO615803.2 - 1 - 2 167 102

View Full Product Details