Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102169288
Entrez Gene ID
102169288
Gene Name
UPF0609 protein C4orf27 homolog
Gene Aliases
C8H4orf27
Chromosome Location
Chr.8: 1541629 - 1565576 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051811.1 XP_017907300.1 7 - 8 993 121
GenBank mRNA JO598774.2 - 1 - 2 300 121
KA311590.1 - 5 - 6 655 121

View Full Product Details